Learn More
Corning™ BioCoat™ Angiogenesis System for Endothelial Cell Invasion
GREENER_CHOICE
Analyze endothelial cell invasion using fluorescence detection in a simplified and reproducible manner
Brand:  Corning™ 354142
 View more versions of this product
Product Code. 11543540
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
HTS insert plates are arrays of individual cell culture inserts connected by a rigid, robotics-friendly holder. This single-unit design makes insert plates ideal for running automated, high throughput drug transport (Caco-2 cells) cell toxicity studies or cell migration and invasion studies.
Corning BioCoat HTS Caco-2 Assay System, PET Membrane Contains specially formulated serum-free medium, culture supplements, sodium butyrate, and the Corning BioCoat Fibrillar Collagen 24-well insert system
TRUSTED_SUSTAINABILITY
Specifications
Specifications
0.3 cm2
3 μm
Endothelial Cell Invasion
1/Pk.
Matrigel™ Matrix
PET
Membrane
5/Pk.
5/Cs.
24
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Corning™ BioCoat™ Angiogenesis System for Endothelial Cell Invasion > 5/Pk