missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ BioCoat™ 96-Well, Anti-Human CD3-Treated, Flat-Bottom Microplate
Color:Clear
Brand: Corning™ 354725
Code : 67
Additional Details : Weight : 0.55000kg
Description
Ideal for applications involving adhesion, growth and differentiation of neuronal and transfected cell lines- Source: rat tail
- TC-treated
- Clear, round well
- Flat bottom
- Non-sterile
- For research use only
Specifications
Standard | |
Clear | |
96 | |
Flat Bottom (F-well) | |
1 | |
2°C to 8°C |
8 x 12 | |
Polystyrene | |
Anti-Human CD3 | |
5 | |
5/Cs. | |
Clear |