missing translation for 'onlineSavingsMsg'
Learn More

Corning™ BioCoat™ 96-Well, Anti-Mouse CD3-Treated, Flat-Bottom Microplate

Color:Clear

Brand:  Corning™ 354720

Code : 67

Additional Details : Weight : 0.50000kg

Product Code. 10794293

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Ideal for applications involving adhesion, growth and differentiation of neuronal and transfected cell lines
  • Source: rat tail
  • TC-treated
  • Clear, round well
  • Flat bottom
  • Non-sterile
  • For research use only
Specifications

Specifications

Standard
Clear
Anti-Mouse CD3
5
5/Cs.
Clear
8 x 12
96
Flat Bottom (F-well)
1
2°C to 8°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers