missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ BioCoat™ 96-Well, T-Cell Activation Control-Treated, Flat-Bottom Microplate
Color:Clear
Brand: Corning™ 354730
Code : 66
Additional Details : Weight : 0.50000kg
Description
Ideal for applications involving adhesion, growth and differentiation of neuronal and transfected cell lines- Source: rat tail
- TC-treated
- Clear, round well
- Flat bottom
- Non-sterile
- For research use only
Specifications
Clear | |
T-Cell Activation Control | |
5 | |
5/Cs. | |
Clear |
96 | |
Flat Bottom (F-well) | |
1 | |
4°C to 30°C |