Learn More
Corning™ BioCoat™ Collagen IV Rectangular Canted Neck Culture Flask
Designed to closely mimic in vivo environments and enhance expression of cell-specific morphology. Corning™ BioCoat™ Collagen IV Rectangular Canted Neck Culture Flask is ideal for applications involving adhesion, growth and differentiation of neuronal and transfected cell lines.
Brand: Corning™ 354528
This item is not returnable.
View return policy
Description
- BioCoat™ provides a superior solution for enhanced cell attachment and growth of a variety of primary cells and transformed cells in serum-free or serum-containing cultures
- Matrix components are applied as an even, optically clear coating
- All lots tested and found negative for bacteria and fungi; cellware is tested for the ability to promote firm attachment of rat cerebellar granule cells
For research use only.
Specifications
Phenolic | |
For adhesion, growth and differentiation of neuronal and transfected cell lines | |
Rectangular | |
Sterile | |
175 cm2 |
750 mL | |
Canted | |
Collagen IV | |
Plug Seal |
Product Content Correction
Product Title
Corning™ BioCoat™ Collagen IV Rectangular Canted Neck Culture Flask > 175 cm2
Spot an opportunity for improvement?Share a Content Correction