missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Falcon™ BioCoat™ 384-Well, Poly-D Lysine-Treated, Flat-Bottom Microplate
Color:Clear; Sterility:Non-sterile; Lid:With Lid
Brand: Falcon™ 356662
Code : 67
Additional Details : Weight : 4.39000kg
Description
Ideal for applications involving adhesion, growth and differentiation of neuronal and transfected cell lines.- Designed to closely mimic in vivo environments and enhance expression of cell-specific morphology
- Matrix components are applied as an even, optically clear coating
- All lots tested and found negative for bacteria and fungi
- Cellware is tested for the ability to promote firm attachment of rat cerebellar granule cells
Specifications
Standard | |
4°C to 30°C | |
5/Pk., 50/Cs. | |
Polystyrene | |
Non-sterile | |
Flat Bottom (F-well) |
With Lid | |
16 x 24 | |
Clear | |
384 | |
Poly-D-Lysine |