Learn More
Molecular Probes™ Biocytin Alexa Fluor™ 488 (Alexa Fluor™ 488 Biocytin, Disodium Salt)
Biocytin Alexa Fluor™ 488 (Alexa Fluor™ 488 Biocytin, Disodium Salt)
Brand:  Molecular Probes™ A12924
313.50 GBP valid until 2025-02-21
Use promo code "24115" to get your promotional price.
Product Code. 11563987
  • £338.00 / 250µg
Explore more special offers
For Research Use Only. All usage must comply with product instructions.
Description
Description
The cell-impermeant, fixable, polar tracer Alexa Fluor™ 488 biocytin combines the green-fluorescent Alexa Fluor™ 488 fluorophore with biotin and an aldehyde-fixable primary amine. Polar tracers are commonly used to investigate cell-cell and cell-liposome fusion as well as membrane permeability and transport through gap junctions or cell uptake during pinocytosis. This water-soluble tracer can be introduced into cells by whole-cell patch clamping, iontophoresis, osmotic lysis of pinocytic vesicles or comparable methods. The biotin allows for detection and subsequent amplification with enzyme-modified streptavidins including horseradish (HRP) and alkaline phosphatase (AP) that can be used with TSA and ELF technologies, respectively.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
250 μg
Alexa Fluor Dyes
Alexa Fluor™
Store at room temperature and protect from light.
Room Temperature