missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Biocytin, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Versatile marker used in anterograde, retrograde and intracellular neuroanatomical investigations and in biotinidase assays. Displays high solubility in aqueous solutions and has a low molecular weight facilitating injection using micropipettes. Can be incorporated with a variety of avidin and streptavidin conjugates for detection by light, fluorescence or electron microscope.
Specifications
Formula Weight | 372.48 |
Percent Purity | >98% |
Quantity | 10 mg |
Chemical Name or Material | Biocytin |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'