missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Biodegradable Detergent

Cleans all types of laboratory glassware, ceramics, plastics, ultrasonic water baths, stainless steel and ferrous metals. Fisherbrand™ Biodegradable Detergent is a very envioronmentally safe cleanser

Brand:  Fisherbrand™ FB70177

Product Code. 11502773

  • £87.40 / Each

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers

Description

Description

A fully biodegradable, phosphate and enzyme free detergent.
  • Removes all biological residues and radioactive contamination
  • Use in concentrations of 2% to 5% in water
  • 5% solution approx. pH 10.8, 2% solution approx. pH 10.3 Suitable for use with all types of laboratory glassware, ceramics, plastics, ultrasonic water baths, stainless steel and ferrous metals.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Biodegradable Detergent
10.8 (5%), 10.3 (2%)
Certificate of analysis
5 L
Liquid
Blue
1 Ea.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Brochures

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Fisherbrand™ Biodegradable Detergent >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.