Learn More
Macherey-Nagel™ BioFix™ Lumi-10 Compact Luminometer
For the measurement of bio and chemical luminescence reactions with constant light emission.
Brand:  Macherey-Nagel™ 940008
Product Code. 10716722
  • £8940.00 / Each
Estimated Shipment: 22-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Compact luminometer for mobile use
    • Ideally suited for use in the laboratory or on the road and can be operated with a power supply as well as rechargeable batteries
  • Incredibly versatile
    • Thanks to its highly sensitive detector (Ultra‐Fast Single Photon Counter) the BioFix™ Lumi‐10 can be used for a variety of applications including, but not limited to, bio toxicity tests, ATP‐ and biomass determinations, reporter‐gene assays, luminescence immunoassays, and NAD(P)H measurements
  • Individually programmable
    • The BioFix™ Lumi‐10 has six individually adjustable measurement protocols and a data memory for up to 2000 results
    • Provides the opportunity for single, multiple and extensive screening measurements
    • Results are optionally displayed in % inhibition, % stimulation or RLU (relative light units)
    • User can set particular measurement parameters such as incubation time or measurement time individually
    • By a previous definition of detection limits, the results can be automatically classified by the device
    • Pre‐programmed test methods are available for the determination of luminescent bacteria toxicity tests and ATP tests
  • Includes manual, rack, cuvettes, and spare adapter
Specifications
Specifications
Backlit graphic display (128 x 64 pixels)
380 nm to 630 nm
50/60 Hz
Manual, rack, cuvettes, and spare adapter, 3 Rechargeable batteries: NiCd R14/C /Baby/UM2 batteries; 1600 mAh, 3 preprogrammed tests, 6 free programmable methods, % inhibition, %stimulation, RLU
RS232 interface for data transfer to the PC or printer
115/230 V
2 kg (with batteries)
CE certified
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Macherey-Nagel™ BioFix™ Lumi-10 Compact Luminometer > Range: 380 nm to 630 nm