missing translation for 'onlineSavingsMsg'
Learn More

Heathrow Scientific™ Biohazard Disposal Bags
GREENER_CHOICE

Product Code. 16320676 Shop All Thermo Scientific CO₂ Incubators Products
Click to view available options
Product Type:
Bag Holder
Benchtop Biohazard Disposal Bag
Capacity (Metric):
2 L
9 L
19 L
40 L
Unit Size:
Each
Pack of 200
Pack of 500
This item is not returnable. View return policy

Product Code. 16320676

Brand: Heathrow Scientific™ HS10321

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Biohazard Disposal Bags for personal or general lab use, multiple sizes available to suit all needs.

  • Biohazard collection bag
  • Biohazard warning symbol and precautions printed in English, Spanish, and French
  • Includes an indicator patch that darkens when autoclaved
  • Autoclavable to 250°F (121°C)
  • Disposable
  • HS10044H bag holder is ONLY for use with HS10320 2 L bag

Specifications

Autoclavable Autoclavable
Capacity (English) 2.4 gal.
Capacity (Metric) 9 L
Color Red
Disposable Single-use
For Use With (Application) Biohazard Disposal
For Use With (Equipment) Autoclave
Material Polypropylene
Sterility Non-sterile
Thickness (Metric) 1.6 mm
No. per Case 200 Pk.
Outer Packaging Box
Thickness (English) 63 mil
Dimensions (L x W) 14 x 19 in.
Dimensions (L x W x H) 14 x 19 x 0.06 in.
Length (English) 14 in.
Length (Metric) 35.6 cm
Width (English) 19 in.
Width (Metric) 48.3 cm
Height (English) 0.06 in.
Height (Metric) 0.15 cm
Weight (English) 0.03 lb.
Weight (Metric) 0.01 kg
Product Type Benchtop Biohazard Disposal Bag
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.