missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bel-Art™ Biohazard Disposal Bags with or without Warning Label
Shop All SP Scienceware Products
Click to view available options
Dimensions (L x W):
24 x 12 in. (61 x 30 cm)
30 x 24 in. (76 x 61 cm)
36 x 24 in. (91 x 61 cm)
Color:
Clear
Translucent
Unit Size:
Pack of 100
Description
Economical, safe disposal of used pipettes, petri dishes, multi-well cell culture plates, cell culture flasks, culture plates, membrane filters, and other biohazard wastes
- These transparent, polypropylene plastic biohazard bags are 0.03 mm (1.5 mil) thick
- High temperature autoclavable at a maximum of 135°C (275°F)
- May also be used as conventional trash can liners
Spezifikation
Spezifikation
| Capacity (Metric) | 35 L |
| Certifications/Compliance | ASTM D1709-98 |
| Closure Type | Zip Tie |
| Design | Biohazard Symbol w/ Sterilization Patch |
| Capacity (English) | 9 gal. |
| Color | Clear |
| Dimensions (L x W) | 30 x 24 in. (76 x 61 cm) |
| Disposable | Single-use |
| Length (Metric) | 76 cm |
| Sterility | Non-sterile |
| Mehr anzeigen |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur