missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ Biohazard Disposal Bags with or without Warning Label

Product Code. p-3207298 Shop All SP Scienceware Products
Click to view available options
Dimensions (L x W):
24 x 12 in. (61 x 30 cm)
30 x 24 in. (76 x 61 cm)
36 x 24 in. (91 x 61 cm)
Color:
Clear
Translucent
Unit Size:
Pack of 100

Product Code. 11392144

Brand: Bel-Art™ F131600005

Please to purchase this item. Need a web account? Register with us today!

Economical, safe disposal of used pipettes, petri dishes, multi-well cell culture plates, cell culture flasks, culture plates, membrane filters, and other biohazard wastes

  • These transparent, polypropylene plastic biohazard bags are 0.03 mm (1.5 mil) thick
  • High temperature autoclavable at a maximum of 135°C (275°F)
  • May also be used as conventional trash can liners

Specifications

Capacity (Metric) 15 L
Certifications/Compliance ASTM D1709-98
Closure Type Zip Tie
Design Non-printed without Sterilization Patch
Capacity (English) 3 gal.
Color Translucent
Dimensions (L x W) 24 x 12 in. (61 x 30 cm)
Disposable Single-use
Length (Metric) 61 cm
Sterility Non-sterile
Width (Metric) 30 cm
Autoclavable Autoclavable
For Use With (Application) Disposal of used pipettes, petri dishes, culture plates, membrane filters and more
Width (English) 12 in.
Length (English) 24 in.
Thickness 1.5 mil (0.03 mm)
Material Polypropylene
Product Type Biohazard Bag
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.