missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
Specifications
| Color | Orange |
| Autoclavable | Autoclavable |
| For Use With (Application) | For providing stable support to biohazard sharp objects safety pouch |
| Material | Epoxy-coated Steel Wire |
| Product Type | Sharps Disposal Container |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction