missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Biohazard Specimen Bag
Fisherbrand™ Biohazard Specimen Bag is a three wall biohazard specimen bag designed for safely collecting, storing, and transporting specimen containers
Brand: Fisherbrand™ 12888306_6000
This item is not returnable.
View return policy
Description
- Preprinted with orange and black Biohazard logo and storage instructions
- Contains 2 x 3 in. absorbent pad to reduce contact with up to 3 mL of specimen fluids
Specifications
Zipper | |
2 x 3 in. Absorbent Pad | |
Non-sterile | |
1000 Pk. | |
6 in. |
Clear | |
LDPE | |
6000/Cs. | |
1.80 mil | |
4 in. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction