Learn More
Socorex™ Acura™ Electro 956.08 Micropipette
Lightweight and ergonomically friendly, this electronic multi-channel micropipette comes with a QC certificate, Qualitips™ pipette tip samples and operating instructions. Socorex™ Acura™ Electro 956.08 Micropipette charges quickly and has a long working autonomy.
Brand:  Socorex™ 956.08.200
Product Code. 11849233
  • £937.00 / Each
Estimated Shipment: 18-03-2025
to see stock.
Explore more special offers
Add to basket
Includes: Electronic pipette, individual QC certificate and operating instructions, pipette charging stand, fast charger with AC adapter and cord, additional battery pack, Qualitips pipette tip samples.
Description
Description
  • Exceptional, user-friendly ergonomics.
  • Lightweight.
  • Easy, intuitive, self-teaching program.
  • Large display, reversible right/left reading.
  • Microprocessor-controlled motor.
  • Swift-interchange, rapidly charging, long-working, rechargeable battery pack.
  • Sequential tip ejection.
  • Also fit any single channel volumetric module.
Specifications
Specifications
<±0.9% - <±0.6%
EN ISO 8655, GMP, GLP, NCCLS. IVD 98/79 EEC (in vitro diagnostic medical devices), ED 89/336 EEC, 91/263 EEC, 92/31 EEC, 93/68 EEC, FCCCFR47, part 15 (electromagnetic compatibility), ED 73/23 EEC, 93/68EEC, IEC 61010-1 (electrical safety), ED 2011/65/EC Restriction of Hazardous Substances (RoHS)
8
Electronic Micropipette
Right/Left LCD
1.0μL
1 Each
24 Months
4.8V NiMH
Acura™ 956
Acura™
10 to 200 μL
QC certificate, Qualitips™ pipette tip samples and operating instructions.
<0.6% - <0.25%
<1.5Hours
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products