Learn More
Sartorius™ Biohit™ Proline Single-Channel Mechanical Pipettes
Ideal for universities and colleges or any laboratory seeking cost-efficient, reliable liquid-handling tool. Sartorius™ Biohit™ Proline Single-Channel Mechanical Pipettes are adjustable-range pipettes that can be used with many tip brands.
Brand:  Sartorius™ 720015
260.93 GBP valid until 2024-12-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 10450352
  • £302.00 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Light weight with high accuracy and precision for use by professionals
  • Wide range of pipettes with adjustable or fixed volume ranges
  • Easy volume setting with click stop mechanism
  • Relaxed grip with ergonomic finger support
  • Reduced risk of contamination with Safe-Cone Filters available
  • Compatible with many universal tips, including Optifit Tips and SafetySpace Tips
Proline Plus models feature:
  • Color coding on the pipette
  • Fully autoclavable, with only 3 parts to clear
  • More ergonomic design for lower required pipetting force
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ISO
Autoclavable
Micropipette
Clear volume
Ergonomic
Laboratory
Yes
Optifit
1
0.5 to 10 μL
Safe-Cone
It is ideal for universities and colleges, or any laboratory seeking a cost-efficient liquid handling tool
1 Each
Chemical
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products