Learn More
Thermo Scientific™ BIOLiner™ Swinging-Bucket Rotor
Spin samples at 3,500rpm with the Thermo Scientific™ BIOLiner™ Swinging-Bucket Rotor.
Brand:  Thermo Scientific™ 75003669
 View more versions of this product
Product Code. 10429894
  • £365.31 / Pack of 4
Explore more special offers
Description
Description
Spin samples at 3,500rpm with the Thermo Scientific™ BIOLiner™ Swinging-Bucket Rotor.
Ordering Information:
Order BIOLiner Buckets (75-003-668, set of 4) and Thermo Scientific™ ClickSeal™ Biocontainment Lids (75-003-669, set of four) to complete assembly. Replacement O-rings for lids (75-003-688) and adapters are sold separately. Microplate carriers (50-036-70) and lids (75-003-689) are also available. Contact your sales representative for information.
Compatible with:
Thermo Scientific™ Sorvall™ ST40/40R, SL40/40R, Sorvall Legend™ XT/XTR, Megafuge™ 40, Multifuge™ X3/X3R, Megafuge 40/40R Centrifuge Series
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ClickSeal Biocontainment Lids (Set of 4)
4 x 750 mL
3,500 rpm
10.5 cm
124 mm
90°
98 mm
Swinging Bucket Rotor
2,739 x g
20 cm
13307
Thermo Scientific Sorvall Legend XT/XF, Sorvall ST 40, Multifuge X3/X3F, Megafuge 40 and SL 40/40F Centrifuges
Biocontainment certified by CAMR™, HPA, Porton Down, U.K.
Yes