Learn More
Sperian™ Honeywell™ Bionic Faceshield Visors
Click to view available options
Description:
Complete faceshield unit
Visor only
Lens Material:
Acetate
Polycarbonate
Lens Coating:
Anti-fog
Uncoated
Description
- Extended top of head and chin protection
- Large superior-quality visors for maximum visibility
- Available with either polycarbonate visor for impact resistance or acetate visor for chemical protection
- Comfortable fit with goggles or respirators
- Locking mechanism for more complete safety
- Dielectric design
- Fully adjustable ratchet headgear and headband
- Multiple tilt visor positions
- Extra-soft cushioning on all parts of the shield in contact with the head
- Breathable extra-soft sweatband is removable for easy washing and replacement
- Excellent for the agriculture, chemical, forestry, steels and metals and wind energy industries
- Protects against abrasion, airborne particles, biohazards, chemicals, impact, splashes and more
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Certifications/Compliance | 89/686/EEC, EN170 / EN166 / EN169, ISO 9001 / 2000 |
Lens Coating | Uncoated |
Description | Visor only |
Lens Tint | Clear |
Product Type | Faceshield |
Lens Material | Acetate |