Learn More
Thermo Scientific™ BioProcess Container (BPC), 200 L, for HyPerforma™ Single-Use Mixer DS 300, Bottom drain, irradiated
The 200 L closed-top, irradiated BioProcess Container (BPC) with bottom drain is composed of CX3-9 single web, multi-layer plastic film and that fit inside drums for Thermo Scientific™ HyPerforma Single-Use Mixer DS 300.
Brand: Thermo Scientific™ SH30946.03
Additional Details : Weight : 4.12769kg
Description
The 200 L closed-top, irradiated BioProcess Container (BPC) with bottom drain is composed of CX3-9 single web, multi-layer plastic film and that fit inside drums for Thermo Scientific™ HyPerforma Single-Use Mixer DS 300. Implement industry-proven BPCs and liners in the overall process of shipping and storing your liquid-liquid and powder-liquid bioprocess mixing applications. BPCs and liners come in open- and closed-top design and are manufactured using our proprietary CX5-14 and CX3-9 film that easily fit in every plastic drum size.
Key features of HyPerforma Single-Use Mixer DS 300 BPCs and liners:
- All standard BPCs and liners are designed to fit 'out-of-the-box'
- Available in volume options of 50, 100, and 200 L
- Applicable to open- and closed-top mixing applications
- Powder-liquid and liquid-liquid mixing
- World class CX5-14 and CX3-9 film
- Adaptable to multiple upstream process applications in media and buffer prep
Required accessories:
- HyPerforma Single-Use Mixer DS 300 docking station
- HyPerforma SIngle-Use Mixer DS 300 drums
Recommended for:
- Powder-liquid or liquid-liquid mixing applications in single use
- High-throughput mixing and storage
- Additional configurations are available to suit your application requirements.
Other Available Accessory:
- Reusable Bearing Hub (Cat. No. SV5017777)
Specifications
200 L | |
Packages placed in strong cardboard cartons with product and lot identification | |
Packed and sealed in two independent outer layers in the Class 10,000/ISO Class 7 area | |
HyPerforma™ |