Learn More
Unity Lab Services™ Biosafety Cabinet Start-up Service
Start-up service for biosafety cabinet without prefilter includes certification for NSF 49 in US and EN12469 in Europe. In UK, an appropriate KI test (BSCKI15S or BSCKI18L) must be added. Excludes major assembly.
Brand:  Unity Lab Services™ STARTBSCMVS
418.20 GBP valid until 2024-12-15
Use promo code "21658" to get your promotional price.
Product Code. 17970882
  • £510.00 / 1 unit
Explore more special offers
Description
Description
  • Includes travel and labor unless extensive travel is needed
  • Thermo Fisher Scientific engineer will perform the following tasks, upon arrival, depending on the specific equipment:
    • Check that the equipment has been handled and assembled as per the user manual
    • Check for proper clearances from obstacles, walls, other equipment, doorways, windows, air vents, and source of excessive light, heat, air flow
    • Stabilize, balance, anchor, connect to pre-installed wall mounts (excluding seismic anchors) with the appropriate hardware
    • Ground, and connect to the proper power supply and other equipment suits via proper connections
    • Connect to flammable and pressured gas sources, tanks, water source, with proper fittings, lines, tubing and other hardware
    • Execute manufacturer's user manual instructions
    • Connect factory-installed safety devices such as alarms, back-up systems
    • Ensure that keypad, controls, service menu and user interface are working
    • Setup of user operating profiles
    • Explain operating, maintenance and cleaning requirements and procedures
    • Explain warranty and service plan options
    • Review annual preventative maintenance purchase options and schedule it appropriately
  • The appliance must be unpacked and installed in its place of use, by the customer, before the technician arrives; excludes major assembly (stacking of heavy CO2 incubators and shakers, assembly of biosafety cabinet stands and positioning cabinet on stands)
  • Includes certification of biosafety cabinets for NSF 49 in US and EN12469 in Europe; in UK, an appropriate KI test (BSCKI15S or BSCKI18L) must be added
  • Customer should ensure that site is properly prepared, ahead of time, including supply sources and connections
  • All additional works, as i.e. waiting time or correctional works, will be invoiced additionally according to our recent valid service pricelist
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Biosafety Cabinets
Start-up Service