missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Agilent BioTek BioStack Automated Microplate Stacker - Microplate Stacks
Storage stacks
£1780.00 - £3664.00
Specifications
Product Type | Microplates |
---|---|
For Use With (Equipment) | Bio-tek Biostack automated microplate stacker, unlidded plates ≤14 mm |
For Use With (Application) | Storage |
Product Code | Brand | Includes | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Includes | Price | Quantity & Availability | |||||
15257639
|
Agilent BioTek
7310030 |
2 stacks of 10 microplates |
£1780.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12512747
|
Agilent BioTek
7310008 |
2 stacks of 30 microplates |
£2714.00
Pack of 2 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11563635
|
Agilent BioTek
7310023 |
2 stacks of 50 microplates |
£3664.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- One set
Specifications
Microplates | |
Bio-tek Biostack automated microplate stacker, unlidded plates ≤14 mm | |
Storage |