Learn More
Agilent BioTek BioStack Automated Microplate Stackers
BioTek BioStack microplate stacker is a compact and versatile microplate stacker, compatible with Agilent BioTek washers, dispensers, plate readers, and imaging systems.
POA
Specifications
Operating Temperature Range (Metric) | 18 to 40°C |
---|---|
Hertz | 50/60 Hz |
Voltage | 100/240 V |
Certifications/Compliance | CE and TUV marked. RoHS compliant. |
Humidity | 10 to 85% |
Includes: One instrument integration kit, communication cable, operator manual, microplate pedestal for storage stack loading and unloading, maintenance kit.
Product Code | Brand | Product Type | Plate Type | Holds | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | Plate Type | Holds | Price | Quantity & Availability | |||
17257909
|
Agilent BioTek
BSTCK2WR-SN |
BioStack Microplate Stacker | ANSI/SBS Standard and low profile 96-, 384- and 1536-well plates, Maximum plate height 14.6 mm | 10-, 30- and 50-plate stacks are removable and interchangeable; 96-/384-well plates: Up to 50 plates; 1536-well plates: Up to 75 plates |
POA
Each |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | ||
17277909
|
Agilent BioTek
BSTCK4-SN |
BioStack 4 Microplate Stacker | ANSI/SBS Standard and low profile 96-, 384- and 1536-well plates; 24- and 48-well plates (model dependent) Maximum plate height: 23.2 mm | 10 and 30 plate stacks are removable and interchangeable (50-plate stacks may be used with non-lidded plates only); 96-/384-well plates: Up to 30 plates (with lids); 1536-well plates: Up to 75 plates |
POA
Each |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | ||
17267909
|
Agilent BioTek
BSTCK3WR-SN |
BioStack 3 Microplate Stacker | ANSI/SBS Standard and low profile 96-, 384- and 1536-well plates, Maximum plate height 14.6 mm | 10-, 30- and 50-plate stacks are removable and interchangeable; 96-/384-well plates: Up to 50 plates; 1536-well plates: Up to 75 plates |
POA
Each |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | ||
17287909
|
Agilent BioTek
BSTCKN2-SN |
BioStack NEO Microplate Stacker | ANSI/SBS Standard and low profile 96-, 384- and 1536-well plates, Maximum plate height 14.6 mm | 10-, 30- and 50-plate stacks are removable and interchangeable; 96-/384-well plates: Up to 50 plates; 1536-well plates: Up to 75 plates |
POA
Each |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | ||
Description
BioTek BioStack microplate stacker is a compact and versatile microplate stacker, compatible with Agilent BioTek washers, dispensers, plate readers, and imaging systems. The rapid plate exchange speeds deliver increased throughput and enhanced efficiency and greater productivity
BioStack can accommodate assay workflows for 24- to 1536-well plates. Microscope slides can be stacked for imaging using BioTek's custom slide holders. The BioTek BioStack 4 is a delidding microplate stacker, automating the handling of lidded microplates for sensitive cell-based assays, for complete walkaway automation.
Certifications
CE marked; ETL listed
Specifications
18 to 40°C | |
100/240 V | |
10 to 85% | |
LHC v 2.12 for liquid handling instruments (optional); Gen5 2.01 for readers | |
11.3 kg |
50/60 Hz | |
CE and TUV marked. RoHS compliant. | |
One instrument integration kit; Communication cable; Operator manual; Microplate pedestal for storage stack loading and unloading; Maintenance kit | |
<25 lb. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.