missing translation for 'onlineSavingsMsg'
Learn More

Spectrum Labs™ Biotech CE Dialysis Tubing

Spectrum Labs™ Biotech CE Dialysis Tubing is designed to meet the demands of ongoing and frequent dialysis applications.

Brand:  Spectrum Labs™ 131456

Code : NEW

Additional Details : Weight : 0.10000kg

Product Code. 15330682

  • £555.00 / 10m
Estimated Shipment: 26-09-2024
to see stock.

Description

Description

  • Hydrophilic
  • Symetric porosity
Specifications

Specifications

Dialysis Tubing
20 mm
31 mm flat width
Packaged wet in 0.05% sodium azide
10 m
300 kDa
10 m
3.1 mL/cm
Cellulose ester
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers