missing translation for 'onlineSavingsMsg'
Learn More

Agilent BioTek 405 TS Touch Microplate Washer
GREENER_CHOICE

The BioTek 405 TS washer has an excellent reputation in life science research for outstanding results in many applications.

POA

Specifications

Certifications/Compliance CE and TUV marked. RoHS compliant.
Connections USB (2)
Bottles Optional (4 or 10 L)
Dispense Volume 25 to 3000 μL/well (192-pin manifold); 50 to 3000 μL/well (96-pin manifold)
Plate Type 96- and 384-well, Low Profile and Standard, Solid and Filter Bottom (Option) Plates
View More Specs

Products 12
Product Code Brand Model Includes Price Quantity & Availability  
Product Code Brand Model Includes Price Quantity & Availability  
17237879
GREENER_CHOICE
View Documents Promotion Details
Agilent BioTek
405TSR-SN
405TSR 96-well microplate washing
POA
Each
N/A
Please to purchase this item. Need a web account? Register with us today!
This item requires a price update prior to being ordered.

Request a Price for this item here.

17247879
GREENER_CHOICE
View Documents Promotion Details
Agilent BioTek
405TSRS-SN
405TSRS 96-well microplate washing, Ultrasonic Advantage
POA
Each
N/A
Please to purchase this item. Need a web account? Register with us today!
This item requires a price update prior to being ordered.

Request a Price for this item here.

17267879
GREENER_CHOICE
View Documents Promotion Details
Agilent BioTek
405TSRV-SN
405TSRV Automated Internal 4-Buffer Switching; 96-well microplate washing
POA
Each
N/A
Please to purchase this item. Need a web account? Register with us today!
This item requires a price update prior to being ordered.

Request a Price for this item here.

17277879
GREENER_CHOICE
View Documents Promotion Details
Agilent BioTek
405TSRVS-SN
405TSRVS Automated Internal 4-Buffer Switching; Ultrasonic Advantage; 96-well microplate washing
POA
Each
N/A
Please to purchase this item. Need a web account? Register with us today!
This item requires a price update prior to being ordered.

Request a Price for this item here.

17297879
GREENER_CHOICE
missing translation for 'viewDocuments' Promotion Details
Agilent BioTek
405TSU-SN
405TSU Optimized Cell Washing; 96-/384-well microplate washing
POA
Each
N/A
Please to purchase this item. Need a web account? Register with us today!
This item requires a price update prior to being ordered.

Request a Price for this item here.

17207889
GREENER_CHOICE
missing translation for 'viewDocuments' Promotion Details
Agilent BioTek
405TSUS-SN
405TSUS Optimized Cell Washing; Ultrasonic Advantage; 96-/384-well microplate washing
POA
Each
N/A
Please to purchase this item. Need a web account? Register with us today!
This item requires a price update prior to being ordered.

Request a Price for this item here.

17227889
GREENER_CHOICE
missing translation for 'viewDocuments' Promotion Details
Agilent BioTek
405TSUV-SN
405TSUV Automated Internal 4-Buffer Switching; 96-/384-well microplate washing
POA
Each
N/A
Please to purchase this item. Need a web account? Register with us today!
This item requires a price update prior to being ordered.

Request a Price for this item here.

17237889
GREENER_CHOICE
missing translation for 'viewDocuments' Promotion Details
Agilent BioTek
405TSUVS-SN
405TSUVS Automated Internal 4-Buffer Switching; Ultrasonic Advantage; Optimized Cell Washing; 96-/384-well microplate washing
POA
Each
N/A
Please to purchase this item. Need a web account? Register with us today!
This item requires a price update prior to being ordered.

Request a Price for this item here.

17257879
GREENER_CHOICE
missing translation for 'viewDocuments' Promotion Details
Agilent BioTek
405TSRSQ-SN
405TSRSQ Ultrasonic Advantage; 96-well microplate washing; Verify Technology
POA
Each
N/A
Please to purchase this item. Need a web account? Register with us today!
This item requires a price update prior to being ordered.

Request a Price for this item here.

17287879
GREENER_CHOICE
missing translation for 'viewDocuments' Promotion Details
Agilent BioTek
405TSRVSQ-SN
405TSRVSQ Automated Internal 4-Buffer Switching; Ultrasonic Advantage; 96-well microplate washing; Verify Technology
POA
Each
N/A
Please to purchase this item. Need a web account? Register with us today!
This item requires a price update prior to being ordered.

Request a Price for this item here.

17217889
GREENER_CHOICE
missing translation for 'viewDocuments' Promotion Details
Agilent BioTek
405TSUSQ-SN
405TSUSQ Optimized Cell Washing; Ultrasonic Advantage; 96-/384-well microplate washing; Verify technology
POA
Each
N/A
Please to purchase this item. Need a web account? Register with us today!
This item requires a price update prior to being ordered.

Request a Price for this item here.

17247889
GREENER_CHOICE
missing translation for 'viewDocuments' Promotion Details
Agilent BioTek
405TSUVSQ-SN
405TSUVSQ Automated Internal 4-Buffer Switching; Ultrasonic Advantage; Optimized Cell Washing; 96-/384-well microplate washing; Verify technology
POA
Each
N/A
Please to purchase this item. Need a web account? Register with us today!
This item requires a price update prior to being ordered.

Request a Price for this item here.

Description

Description

The BioTek 405 TS washer has an excellent reputation in life science research for outstanding results in many applications. The 405 TS has a unique set of features designed to efficiently wash cell-based assays, microsphere-based assays, and ELISA applications. Programming and operation via the touchscreen interface is intuitive and simple.

The patented Verify technology automatically checks for manifold tube blockages before a wash cycle, and Ultrasonic Advantage automatically cleans any blocked manifold tubes. These functions save time and eliminate frustration while ensuring successful results.

 

Must be purchased together with required dispense/waste system accessory for use. Please contact your Fisher Sales Representative for more information.

Specifications

Specifications

CE and TUV marked. RoHS compliant.
Optional (4 or 10 L)
96- and 384-well, Low Profile and Standard, Solid and Filter Bottom (Option) Plates
Programmable in minutes and seconds, up to 60 min.
With internal buffer switching - 16.5 kg; Without - 13.5 kg
25 to 3000 μL/well
Internal positive displacement pump
14 in.
10 in.
17 in.
High Resolution LED Backlit Touch Screen
100/240 V
USB (2)
25 to 3000 μL/well (192-pin manifold); 50 to 3000 μL/well (96-pin manifold)
≤2 μL/well
Microplate Washer
1 to 250
LED
50/60 Hz
35.6 cm
25.4 cm
43.2 cm
With internal buffer switching - 36 lb.; Without - 30 lb.
Videos
SDS
Documents

Documents

Certificates