Learn More
Agilent BioTek Sodium Fluorescein Powder
1 mg vial
Brand: Agilent BioTek 98155
Additional Details : CAS Number : 518-47-8 Weight : 0.01000kg
Description
- Sodium Fluorescein Powder (1 mg vial)
- Used for FLx800 and Synergy fluorescence qualification tests
- For use with Agilent BioTek Microplate Washers and Readers
Specifications
Sodium Fluorescein Powder | |
1 mg | |
Agilent BioTek Microplate Washer and Reader Instruments |