Learn More
Agilent BioTek Liquid Handling Control Software
Full PC control provided by LHC Software leads to increased convenience when programming important assay-specific protocol requirements in a familiar Windows environment. Fisher Scientific - Liquid Handling Control Software is ideal for use with 405TM Touch Washer, 405TM LS Washer, ELx405TM Select Deep Well Washer, EL406TM Washer Dispenser and MultiFloTMFX Dispenser.
Brand: Agilent BioTek LHC2SECURE
Includes: Software on CD, installation guide and 5 licenses
Description
- Intuitive StepWise protocol creation with an unlimited number of methods included for ultimate flexibility
- Automatic protocol validation for error-free assurance
- Dual mode control to create and run protocols from PC or download to instrument keypad
- Security enabled protection for downloaded protocols with LHC2 SECURE
- Printed protocol reports and automatic protocol history
- Custom maintenance reminders for continued instrument performance in busy laboratories
- Multiple instruments controlled in parallel, including BioStack integrations for unattended batch processing
Compliance
United States Food and Drug Administration 21 CFR Part 11 regulation (LHC Secure Edition)
Specifications
21 CFR Part 11 | |
Software on CD, installation guide and 5 licenses | |
BioTek 405 Touch Washer, 405 LS Washer, ELx405 Select Deep Well Washer, EL406 Washer Dispenser and MultiFlo Dispenser | |
Liquid Handling Control Software |