Learn More
Agilent BioTek Washer Biomagnetic Separation Carrier Accessories
Shop All Agilent BioTek Products
Click to view available options
Type:
384-Well Ring Magnet
96-Well Ring Magnet
Unit Size:
Each
Description
- Magnet ring
- External
- For insertion into washer biomagnetic separation carriers
- Beads pulled to 4-zone ring at well bottom
Specifications
Specifications
| Type | 96-Well Ring Magnet |
| For Use With (Equipment) | 405 LS Microplate Washer, 405 Touch Microplate Washer, EL406 Microplate Washer, ELx 405 Multiflo FX Microplate Washer |