Learn More
Tocris Bioscience™ Biotin-11-dUTP
Biotin-labeled dUTP
Brand: Tocris Bioscience™ 6571/200UL
Additional Details : Weight : 0.00970kg
Description
Biotin-11-dUTP is a biotin-labeled dUTP. Commonly used in PCR, nick translation, and reverse transcription. Also frequently used in apoptosis assays. Biotin-labeled DNA may be detected with streptavidin-HRP conjugates.Specifications
2 '-Deoxy-5-[(1 E)-3-[[6-[[5-[(3 AS,4 S,6 aR)-hexahydro-2-oxo-1 H-thieno[3,4-d]imidazol-4-yl]-1-oxopentyl]amino]-1-oxohexyl]amino]-1-propen-1-yl]uridine 5 '-(tetra(triethylammonium) triphosphate) | |
Buffers, Chelators and Reagents | |
95% |
200 μL | |
C28H45N6O17P3S0.4(C2H5)3N |