Learn More
IBA Lifesciences Biotin
![GREENER_CHOICE](/content/dam/fishersci/glyphs/green_glyph.png)
Click to view available options
Quantity:
2 g
5 g
Description
- Biotin is a water-soluble B-complex vitamin, also known as vitamin B7 or vitamin H. It binds to the biotin-binding pockets of streptavidin, Strep-Tactin™, and Strep-Tactin™XT with different affinities.
- It allows the efficient elution of Strep-tag™II or Twin-Strep-tag™ fusion proteins from Strep-Tactin™XT, while maintaining the reusability of the resin.
- It can be used for preparation of Strep-Tactin™XT elution buffer, Buffer BXT.
- In addition, biotin can be used for elution of biotinylated proteins from Strep-Tactin™, where it binds irreversibly to the resin - Strep-Tactin™ cannot be regenerated in this kind of application.
Specifications
Quantity | 5 g |