missing translation for 'onlineSavingsMsg'
Learn More
(+)-Biotin 4-nitrophenyl ester, 98%, Thermo Scientific™ Product Code.: 15437378

(+)-Biotin 4-nitrophenyl ester, 98%, Thermo Scientific™

Product Code. 15437378
100 mg
Click to view available options
Quantity:
100 mg
This item is not returnable. View return policy

Product Code. 15437378

Brand: Thermo Scientific Alfa Aesar H60873.MC

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 33755-53-2
Melting Point 163°C to 165°C
Molecular Formula C16H19N3O5S
MDL Number MFCD15141972
Quantity 100 mg
Beilstein 5795805
Synonym +-biotin 4-nitrophenyl ester, d-biotin p-nitrophenyl ester, biotinyl-4-nitrophenyl ester, bnp, d-biotin 4-nitrophenyl ester, biotin-p-nitrophenyl ester, 4-nitrophenyl 5-3as,4s,6ar-2-oxo-hexahydrothieno 3,4-d imidazol-4-yl pentanoate, biotin p-nitrophenyl ester, 4-nitrophenyl +-biotinate, +-biotin p-nitrophenyl ester, 4-nitrophenyl 5-3as,4s,6ar-2-oxohexahydro-1h-thieno 3,4-d imidazol-4-yl pentanoate
InChI Key YUDNXDTXQPYKCA-YDHLFZDLSA-N
Optical Rotation +51° (c=2 in DMF:AcOH)
SMILES [O-][N+](=O)C1=CC=C(OC(=O)CCCC[C@@H]2SC[C@@H]3NC(=O)N[C@H]23)C=C1
IUPAC Name 4-nitrophenyl 5-[(3aS,4S,6aR)-2-oxo-hexahydro-1H-thieno[3,4-d]imidazol-4-yl]pentanoate
Molecular Weight (g/mol) 365.40
PubChem CID 83848
Formula Weight 365.4
Percent Purity 98%
Chemical Name or Material (+)-Biotin 4-nitrophenyl ester
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
(+)-Biotin 4-nitrophenyl ester, 98%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.