missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Biotin-DADOO

Product Code. p-200104427
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 16418778

Brand: MedChemExpress HYD0980

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

MedChemExpress Biotin-DADOO is a biotinylation reagent, which can be used to synthesize a biotin-estradiol conjugate (i.e., biotin-DADOO-estradiol) to develop a direct, broad range enzyme immunoassay to measure plasma estradiol concentrations.

Specifications

Quantity 10 mg
Product Type Biochemical Assay Reagents
Content And Storage Powder -20°C 3 years, 4°C 2 years ∣ In solvent -80°C 6 months, -20°C 1 month
Shipping Condition Room temperature
CAS 138529-46-1
Molecular Weight (g/mol) 374.5 g/mol
Solubility DMSO : 100 mg/mL (267.02 mM; Need ultrasonic) ∣ H2O : ≥ 100 mg/mL (267.02 mM)
Synonym Biotinyl-3,6-dioxaoctanediamine ∣ EZ-Link Amine-PEO2-Biotin
Target HBV
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.