Learn More
Invitrogen™ Biotin-HRP (biotinylated peroxidase)
Biotin-HRP (biotinylated peroxidase)
Brand: Invitrogen™ 432040
Code : 50
Additional Details : Weight : 0.02100kg
Description
Reactivity: Reacts with streptavidin and avidin conjugates.Specifications
Wet Ice | |
Store in refrigerator at 2°C to 8°C | |
Liquid | |
HRP (Horseradish Peroxidase) |
Safety and Handling
- BIOTINYLATED PEROXIDASE
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
Hazard Statement
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- H319-Causes serious eye irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P272-Contaminated work clothing should not be allowed out of the workplace.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P362+P364-Take off contaminated clothing and wash it before reuse.
Supplemental information
- MIXTURE LIST-Contains : 2-methyl-3(2H)-isothiazolone
For Research Use Only. Not for use in diagnostic procedures.