missing translation for 'onlineSavingsMsg'
Learn More

Biotin, MedChemExpress

Product Code. p-200097753
Encompass_Preferred
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 16460968

Brand: Medchem Express HYB0511

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Biotin (Vitamin B7) is a water-soluble B vitamin and serves as a coenzyme for five carboxylases in humans, involved in the synthesis of fatty acids, isoleucine, and valine, and in gluconeogenesis. Biotin is necessary for cell growth, the production of fatty acids, and the metabolism of fats and amino acids.

Biotin (Vitamin B7) is a water-soluble B vitamin and serves as a coenzyme for five carboxylases in humans, involved in the synthesis of fatty acids, isoleucine, and valine, and in gluconeogenesis. Biotin is necessary for cell growth, the production of fatty acids, and the metabolism of fats and amino acids.

Specifications

Chemical Name or Material Biotin
CAS 58-85-5
Color White
Purity Grade Notes Research
Molecular Formula C10H16N2O3S
Quantity 5 g
Synonym Vitamin B7 Vitamin H D-Biotin
Solubility Information DMSO : 100 mg/mL (409.32 mM; Need ultrasonic) ∣H2O : 1 mg/mL (4.09 mM; Need ultrasonic)
SMILES O=C(O)CCCC[C@@H]1SC[C@]([C@]1([H])N2)([H])NC2=O
Molecular Weight (g/mol) 244.31
Formula Weight 244.31
Percent Purity 98.0%
Grade Research
Physical Form Powder
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.