missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals (+)-Biotin N-hydroxysuccinimide ester, 98% Product Code.: 11986841

Thermo Scientific Chemicals (+)-Biotin N-hydroxysuccinimide ester, 98%

Product Code. 11986841
250 mg, Glass bottle
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 11986841

Brand: Thermo Scientific Chemicals 439302500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35013-72-0
Molecular Formula C14H19N3O5S
Molecular Weight (g/mol) 341.39
InChI Key YMXHPSHLTSZXKH-RVBZMBCESA-N
Synonym +-biotin n-hydroxysuccinimide ester, biotin-nhs, biotin nhs, 2,5-dioxopyrrolidin-1-yl 5-3as,4s,6ar-2-oxohexahydro-1h-thieno 3,4-d imidazol-4-yl pentanoate, n-hydroxysuccinimidobiotin, d-biotinyl-osu, biotin-osu, n-succinimidyl d-biotinate, bhse, biotin n-hydroxysuccinimide ester
PubChem CID 6710714
IUPAC Name (2,5-dioxopyrrolidin-1-yl) 5-[(3aS,4S,6aR)-2-oxo-1,3,3a,4,6,6a-hexahydrothieno[3,4-d]imidazol-4-yl]pentanoate
SMILES C1CC(=O)N(C1=O)OC(=O)CCCCC2C3C(CS2)NC(=O)N3

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 202.3°C to 202.7°C
Color White
Elemental Analysis 48.03 to 50.49% C, 12.00 to 12.62% N
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 250 mg
Formula Weight 341.39
Percent Purity 98%
Physical Form Powder
Chemical Name or Material (+)-Biotin N-hydroxysuccinimide ester
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number345399 Certificate TypeCertificate of Analysis Date04/09/2024 Catalog Number439300010,439302500
Lot Number341113 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number439300010,439302500
Lot Number344867 Certificate TypeCertificate of Analysis Date14/09/2022 Catalog Number439300010,439302500
Lot Number344891 Certificate TypeCertificate of Analysis Date14/09/2022 Catalog Number439300010,439302500
Lot Number344220 Certificate TypeCertificate of Analysis Date29/11/2021 Catalog Number439300010,439302500
Product Identifier
  • (+)-Biotin N-hydroxysuccinimide ester, 98%

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals (+)-Biotin N-hydroxysuccinimide ester, 98% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.