missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Biotin-Substance P
For cellular and molecular biology applications
£268.00 - £407.00
Specifications
Purification Method | HPLC |
---|---|
Content And Storage | -30°C to -10°C |
Format | Solid |
Molecular Weight (g/mol) | 1573.94 |
Product Type | Biotin-Substance P |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15448099
|
Thermo Scientific Chemicals
|
10 mg |
£407.00
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15438099
|
Thermo Scientific Chemicals
J66653.#0 |
5 mg |
£268.00
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
HPLC | |
Solid | |
Biotin-Substance P | |
Biotin-Arg-Pro-Lys-Pro-Gln-Gln-Phe-Phe-Gly-Leu-Met-NH2 |
-30°C to -10°C | |
1573.94 | |
RUO - research use only |
RUO – Research Use Only