missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Biotin (Vitamin B7 or Vitamin H) Antibody (SPM375), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-32812-0.02mg
Additional Details : Weight : 0.00970kg
Description
Biotin (Vitamin B7 or Vitamin H) Monoclonal antibody specifically detects Biotin (Vitamin B7 or Vitamin H) in Nonspecies specific samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), In-situ HybridizationSpecifications
Biotin (Vitamin B7 or Vitamin H) | |
Monoclonal | |
0.2 mg/mL | |
Western Blot 0.5-1ug/mL, Flow Cytometry 1-2 ug/million cells, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 1-2 ug/mL, Immunohistochemistry-Paraffin 1-2 ug/mL, In-situ Hybridization | |
58-85-5, coenzyme R, d-biotin, Vitamin B7, vitamin H | |
Biotinylated sheep immunoglobulin | |
0.02 mg | |
Primary | |
Store at 4C. | |
IgG1 κ |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin), In Situ Hybridization (ISH) | |
SPM375 | |
Unconjugated | |
10 mM PBS with 0.05% BSA | |
Mouse | |
Protein A or G purified | |
RUO | |
Non-species specific | |
Purified |