Learn More
Molecular Probes™ Biotin-X Cadaverine (5-(((N-(Biotinoyl)Amino)Hexanoyl)Amino)Pentylamine, Trifluoroacetic Acid Salt)
Biotin-X Cadaverine (5-(((N-(Biotinoyl)Amino)Hexanoyl)Amino)Pentylamine, Trifluoroacetic Acid Salt)
Brand: Molecular Probes™ B1596
Code : 50
Additional Details : Weight : 0.00525kg
Description
The primary aliphatic amine of biotin-X cadaverine can be reversibly coupled to aldehydes and ketones to form a Schiff base - which can be reduced to a stable amine derivative by sodium borohydride (NaBH4) or sodium cyanoborohydride (NaCNH3) to form new biotinylated probes. Carboxylic acids of proteins and other water-soluble biopolymers can be coupled to this molecule in aqueous solution using water-soluble carbodiimides such as EDAC (E2247).Specifications
Biotin-X Cadaverine | |
Store at room temperature. | |
Carboxylic Acid, Ketone, Aldehyde | |
Biotin-X |
10 mg | |
Room Temperature | |
Amine, Cadaverine | |
Biotin & Other Haptens |
For Research Use Only. Not for use in diagnostic procedures.