missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Biotin XX Tyramide SuperBoost™ Kit, Goat anti-rabbit IgG
Biotin XX Tyramide SuperBoost™ Kit features biotin XX tyramide with poly-HRP-conjugated goat anti-rabbit IgG secondary antibody.
Brand: Invitrogen™ B40921
Additional Details : Weight : 0.17880kg
Description
SuperBoost™ tyramide signal amplification is most sensitive method for detection of low abundant targets in multiplexable fluorescent immunocytochemistry (ICC), immunohistochemistry (IHC ), and in situ hybridization (ISH).Features:
- High sensitivity for detection of low-level or hard-to-detect targets by fluorescent imaging
- Simple protocol and detection using standard filters
- Suitable for high-resolution multiplex images, co-label with DAPI, secondary antibodies, and other SuperBoost kits
- Requires 10-100 times less primary antibody then standard ICC/IHC/ISH experiments
Benefits:
- Poly-HRP enhancement: Several HRP enzymes are conjugated with short polymers, enhancing signal by several fold over regular HRP systems
- Reaction stop solution: Include an HRP stop solution to halt the HRP reaction
- Reduction of background: Include blockers for elimination or reduction of endogenous peroxidase and fluorescent background signals
Shelf Life:
- Stable for 6 months after receiving
Specifications
Biotin-XX | |
Blocking buffer (1X): 22.5mL, Poly-HRP-conjugated goat anti-rabbit secondary antibody (1X): 22.5mL, Biotin-XX tyramide reagent, Hydrogen peroxide (stabilized 3% solution): 28.5mL, Reaction buffer (20X): 6mL, Reaction stop solution: 8mg, Dimethylsulfoxide | |
SuperBoost™ Kit |
2°C to 8°C | |
150 slides |
For Research Use Only. Not for use in diagnostic procedures.