missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Biotrak™ Human MMP-1 Assay

Suitable for use with cell culture supernatant, serum and plasma samples

Brand:  Cytiva RPN2610

Product Code. 10438454

This item is not returnable. View return policy

Description

Description

  • Specific for total MMP-1: recognizes pro MMP-1, active MMP-1 and MMP-1/TIMP-1 complex
  • Range: 6.25-100ng/mL
  • Sensitivity: 1.7ng/mL
  • 5.5 hour protocol

TRUSTED_SUSTAINABILITY
Specifications

Specifications

MMP-1
Human
Each
1.7 ng/mL
-15°C to -30°C
Human MMP-1 ELISA
6.25 to 100ng/mL
Microplate (12 x 8-well strip coated with Mouse Anti-MMP-1), Assay Buffer 1, Assay Buffer 2, 1mL Human MMP-1 standard (recombinant), Rabbit Anti-MMP-1 Antibody (lyophilised), donkey Anti-Rabbit HRP (lyophilised), Wash Buffer, TMB Substrate
Cell Culture Supernatant, Serum, Plasma
5.5 hr.
Measure MMP Protein levels
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • MMP-1,Human, Assay

Hazard Statement
  • EUH210-Safety data sheet available on request.

Supplemental information
  • UNKNOWN ACUTE-(RPN2610:)100 per cent of the mixture consists of component(s) of unknown acute toxicity.
  • UNKNOWN AQUATIC-Contains (RPN2610:)100% of components with unknown hazards to the aquatic environment.

SDS
Documents

Documents

Certificates
Special Offers

Special Offers