Learn More
Cytiva BioTrak™ Human TNFα Easy ELISA
Suitable for use with cell culture supernatant, serum, plasma and amniotic fluid samples
Brand: Cytiva RPN5967
Code : GE
Additional Details : CAS Number : 53123-88-9 Weight : 0.35000kg
Description
- Range: 7.8 to 500pg/mL
- Sensitivity: 2.5pg/mL
- Sample: 100μL 210 min. protocol
Specifications
TNF-alpha | |
Certificate of Analysis | |
Each | |
50 μL | |
210 min. | |
Quantitative detection of Human Tumor Necrosis Factor a in cell culture supernatants, Human Serum, Plasma, Urine, Synovial fluid, Amniotic fluid or other body fluid |
7.8 to 500pg/mL | |
96-well microplate coated with Mab (murine), 2 TNFa standard curve, 1 bottle x 25mL Wash Buffer 20X concentrate, 1 vial x 15mL Substrate solution, 1 vial 12mL sample diluent, 1 vial x 15mL stop solution, 1 adhesive plate cover | |
Cell Culture Supernatant, Serum, Plasma | |
1.65 pg/mL | |
-20°C | |
Human TNFalpha easy ELISA |