Learn More
Cytiva Biotrak™ Thromboxane B2 assay
A rapid and sensitive non-isotopic immunoassay method for the determination of thromboxane B2 in plasma and serum samples
Brand: Cytiva RPN220
Code : GH
Additional Details : CAS Number : 55965-84-9 Weight : 0.45200kg
Description
- Range: 10-1280pg/mL (0.5-64pg/well)
- High sensitivity: 0.18pg/well, 3.6pg/mL
- Two-hour protocol (15-20 hours for maximum sensitivity
Specifications
Thromboxane B2 | |
Microplate (12 x 8-well strip coated with donkey Anti-Rabbit IgG), Thromboxane B2 peroxidase Conjugate (lyophilized), 2.56ng standard (Thromboxane B2), antiserum (Rabbit Anti-thromboxane B2, lyophilized), TMB Substrate, Assay Buffer concentrate, Wash Buffer concentrate | |
50 μL | |
2 hr. | |
ELISA |
10 to 1280pg/mL | |
Each | |
1.18 to 3.6 pg/mL | |
2°C to 8°C | |
Thromboxane B2 assay |