Learn More
Welch Ilmvac™ Biovac 106 Aspiration System
Use this aspirator for aspiration and disposal of liquids in biochemistry, microbiology and cell culture applications. Welch Ilmvac™ Biovac 106 Aspiration Systems are idea for precise aspiration of chemical, biological and medical liquids.
Brand: Welch Ilmvac™ 112037
This item is not returnable.
View return policy
Description
- Aspiration and filtration system
- Includes handheld HandVac pipettor
- Autoclavable collection receiver
- Automatic flow stop and in-line hydrophobic filter
- Available with 2 liter glass or 4 liter PP collection receiver
Specifications
115/230 V 50/60 Hz | |
8 kg | |
Aspiration System |
Biovac 106 Aspiration System | |
15 L/min. | |
<100 mbar |
SAS035 112012 Biovac 106
File size: 956 kB
Product Content Correction
Product Title
Welch Ilmvac™ Biovac 106 Aspiration System > Welch Ilmvac™
Spot an opportunity for improvement?Share a Content Correction