missing translation for 'onlineSavingsMsg'
Learn More

Biochrom Biowave 3

The Biowave 3 offers all the capabilities of a research grade spectrophotometer with a robust design and a reasonable cost make it great for students.

Brand:  Biochrom 80-3007-35


Additional Details : Weight : 3.00000kg

 View more versions of this product

Product Code. 17213064

  • £6385.00 / Each
Estimated Shipment: 18-09-2024
to see stock.

Description

Description

  • UV-Vis Spectrophotometer with a 5 nm bandwidth
  • Features a USB port for data collection
  • Pre-programmed life science methodologies, including nucleic acid quantification (DNA, RNA and oligonucleotides), protein assays, and for cell culture density (OD600) measurements
  • Nucleic acid scans for purity checking
  • Low volume capability for use with microvolume cuvettes
  • Novel optics with a Xenon source for long lamp lifetime and low running costs
  • Choose to include a built-in serial printer, Bluetooth connectivity, or both
  • Lightweight and stackable for storage
  • Spectrophotometer has a 2 year warranty and the Xenon flash lamp has a 3 year warranty
  • IQ/OQ available on request
Specifications

Specifications

15 mm
390 mm
Graphic LCD
Xenon Flash
5 nm
100/115/220/230 V
UV/Vis Spectrophotometry
3.9 in.
Spectrophotometer, Bluetooth Connectivity, Built-in Serial Printer
Flat Grating
±1.3% or ±0.008 A (Whichever is Greater at 546 nm)
±0.002 A to 0.5 A (at 546 nm)
Round
±2 nm
6.6 lb.
10 in.
15.3 in.
Twin CMOS Array
English, French, German, Spanish, Italian, Chinese, Japanese
Biowave 3
UV-Vis Spectrophotometer
190 to 1100 nm
Microvolume Cuvettes
100 mm
Large Display, Button UI, USB Port, PC via PVC Software, Bluetooth Connectivity
±0.005 peak to peak ±0.002 RMS
-0.3 to 2.5 A, 0.3 to 199 %T
Yes
>0.5 %T 340 nm
±1 nm
3 kg
260 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers