missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Biochrom Biowave 3
The Biowave 3 offers all the capabilities of a research grade spectrophotometer with a robust design and a reasonable cost make it great for students.
Brand: Biochrom 80-3007-33
Description
- UV-Vis Spectrophotometer with a 5 nm bandwidth
- Features a USB port for data collection
- Pre-programmed life science methodologies, including nucleic acid quantification (DNA, RNA and oligonucleotides), protein assays, and for cell culture density (OD600) measurements
- Nucleic acid scans for purity checking
- Low volume capability for use with microvolume cuvettes
- Novel optics with a Xenon source for long lamp lifetime and low running costs
- Choose to include a built-in serial printer, Bluetooth connectivity, or both
- Lightweight and stackable for storage
- Spectrophotometer has a 2 year warranty and the Xenon flash lamp has a 3 year warranty
- IQ/OQ available on request
Specifications
15 mm | |
390 mm | |
Graphic LCD | |
Xenon Flash | |
5 nm | |
100/115/220/230 V | |
UV/Vis Spectrophotometry | |
3.9 in. | |
Spectrophotometer, Built-in Serial Printer | |
Flat Grating | |
±1.3% or ±0.008 A (Whichever is Greater at 546 nm) | |
±0.002 A to 0.5 A (at 546 nm) | |
Round | |
±2 nm | |
6.6 lb. | |
10 in. |
15.3 in. | |
Twin CMOS Array | |
English, French, German, Spanish, Italian, Chinese, Japanese | |
Biowave 3 | |
UV-Vis Spectrophotometer | |
190 to 1100 nm | |
Microvolume Cuvettes | |
100 mm | |
Large Display, Button UI, USB Port, PC via PVC Software | |
±0.005 peak to peak ±0.002 RMS | |
-0.3 to 2.5 A, 0.3 to 199 %T | |
Yes | |
>0.5 %T 340 nm | |
±1 nm | |
3 kg | |
260 mm |