missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Biochrom Biowave 3+
The Biowave 3+ offers all the capabilities of a research grade UV/Vis spectrophotometer. Expect exceptionally high resolution with the 3 nm bandwidth. The robust design and reasonable cost make it great for students.
£6380.00 - £6795.00
Specifications
Beam Center Height | 15 mm |
---|---|
Depth (English) | 15.3 in. |
Depth (Metric) | 390 mm |
Detector Type | Twin CMOS Array |
Display | Graphic LCD |
Product Code | Brand | Includes | Interface | Printer Included | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Includes | Interface | Printer Included | Price | Quantity & Availability | |||
17120509
|
Biochrom
80-3007-37 |
Spectrophotometer | Large Display, Button UI, USB Port, PC via PVC Software | No |
£6380.00
Each |
Estimated Shipment: 20-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17223064
|
Biochrom
80-3007-38 |
Spectrophotometer, Built-in Serial Printer | Large Display, Button UI, USB Port, PC via PVC Software | Yes |
£6465.00
Each |
Estimated Shipment: 15-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17233064
|
Biochrom
80-3007-39 |
Spectrophotometer, Bluetooth Connectivity | Large Display, Button UI, USB Port, PC via PVC Software, Bluetooth Connectivity | No |
£6645.00
Each |
Estimated Shipment: 15-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17243064
|
Biochrom
80-3007-40 |
Spectrophotometer, Bluetooth Connectivity, Built-in Serial Printer | Large Display, Button UI, USB Port, PC via PVC Software, Bluetooth Connectivity | Yes |
£6795.00
Each |
Estimated Shipment: 15-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
Description
- UV-Vis Spectrophotometer with a 3 nm bandwidth
- Features a USB port for data collection
- Pre-programmed life science methodologies, including nucleic acid quantification (DNA, RNA and oligonucleotides), protein assays, and for cell culture density (OD600) measurements
- Nucleic acid scans for purity checking
- Low volume capability for use with microvolume cuvettes
- Novel optics with a Xenon source for long lamp lifetime and low running costs
- Choose to include a built-in serial printer, Bluetooth connectivity, or both
- Lightweight and stackable for storage
- Spectrophotometer has a 2 year warranty and the Xenon flash lamp has a 3 year warranty
- IQ/OQ available on request
Specifications
15 mm | |
390 mm | |
Graphic LCD | |
Xenon Flash | |
100/115/220/230 V | |
10x10mm cuvettes | |
100 mm | |
±0.005 peak to peak ±0.002 RMS | |
-0.3 to 2.5 A, 0.3 to 199 %T | |
Round | |
±2 nm | |
6.6 lb. | |
10 in. |
15.3 in. | |
Twin CMOS Array | |
English, French, German, Spanish, Italian, Chinese, Japanese | |
3 nm | |
190 to 1100 nm | |
3.9 in. | |
Flat Grating | |
±1.3% or ±0.008 A (Whichever is Greater at 546 nm) | |
±0.002 A to 0.5 A (at 546 nm) | |
<0.5 %T 340 nm | |
±1 nm | |
3 kg | |
260 mm |