missing translation for 'onlineSavingsMsg'
Learn More

Biphenyl-2-boronic acid pinacol ester, 97%

Artikelnummer. 15445037
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
25 g
missing translation for 'unitSize'
1g
25g
5g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Artikelnummer. 15445037

missing translation for 'mfr': Thermo Scientific Alfa Aesar H32320.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

CAS: 914675-52-8 | C18H21BO2 | 280.174 g/mol

Biphenyl-2-boronic acid pinacol ester, is used as a main benefits of the use of boronic esters in Suzuki-Miyaura cross-couplings- Enabling the coupling of organoboronic acids that are sensitive to hydrolytic deboronation. It includes other additional application such as a number of chiral boronic esters, such as the ones illustrated hereafter, have been used as inducers in stereoselective reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Biphenyl-2-boronic acid pinacol ester, is used as a main benefits of the use of boronic esters in Suzuki-Miyaura cross-couplings- Enabling the coupling of organoboronic acids that are sensitive to hydrolytic deboronation. It includes other additional application such as a number of chiral boronic esters, such as the ones illustrated hereafter, have been used as inducers in stereoselective reactions.

Solubility
Insoluble in water. Soluble in organic solvents.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 914675-52-8
Molecular Formula C18H21BO2
Molecular Weight (g/mol) 280.174
MDL Number MFCD14636459
InChI Key WCXWQEUBHZKNMQ-UHFFFAOYSA-N
Synonym biphenyl-2-boronic acid pinacol ester, 2-1,1'-biphenyl-2-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-2-biphenylyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
PubChem CID 59207619
IUPAC Name 4,4,5,5-tetramethyl-2-(2-phenylphenyl)-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC=CC=C2C3=CC=CC=C3

Spezifikation

Melting Point 76°C to 79°C
Quantity 5 g
Solubility Information Insoluble in water. Soluble in organic solvents.
Formula Weight 280.17
Percent Purity 97%
Chemical Name or Material Biphenyl-2-boronic acid pinacol ester

RUO – Research Use Only

Berichtigung von Produktinhalten

Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.

Name des Produkts

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt