missing translation for 'onlineSavingsMsg'
Learn More
Biphenyl-4,4'-dicarbonitrile, 98%, Thermo Scientific Chemicals Product Code.: 11322166

Biphenyl-4,4'-dicarbonitrile, 98%, Thermo Scientific Chemicals

Product Code. 11322166
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11322166

Brand: Thermo Scientific Alfa Aesar L03450.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Biphenyl-4,4'-dicarbonitrile is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Biphenyl-4,4′-dicarbonitrile is used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1591-30-6
Molecular Formula C14H8N2
Molecular Weight (g/mol) 204.232
MDL Number MFCD00013805
InChI Key KAXYYLCSSXFXKR-UHFFFAOYSA-N
Synonym 4,4'-biphenyldicarbonitrile, 1,1'-biphenyl-4,4'-dicarbonitrile, 4,4'-dicyanobiphenyl, biphenyl-4,4'-dicarbonitrile, 4,4'-dicyanodiphenyl, 4,4'-diphenyldicarbonitrile, 4-4-cyanophenyl benzonitrile, 4-4-cyanophenyl benzenecarbonitrile
PubChem CID 15321
IUPAC Name 4-(4-cyanophenyl)benzonitrile
SMILES C1=CC(=CC=C1C#N)C2=CC=C(C=C2)C#N

Specifications

Melting Point 232°C to 236°C
Quantity 1 g
UN Number UN3439
Beilstein 1869917
Solubility Information Insoluble in water.
Formula Weight 204.23
Percent Purity 98%
Chemical Name or Material Biphenyl-4,4'-dicarbonitrile

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10219288 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberL03450.06,L03450.03
Lot NumberU05A017 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL03450.06,L03450.03
Lot Number10164026 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL03450.06,L03450.03
Lot Number10152961 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL03450.06,L03450.03
Lot Number10219288 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL03450.06,L03450.03

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Biphenyl-4,4'-dicarbonitrile, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.