missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Biphenyl-4-boronic acid pinacol ester, 97%, Thermo Scientific Chemicals
£44.70 - £188.00
Chemical Identifiers
CAS | 144432-80-4 |
---|---|
Molecular Formula | C18H21BO2 |
Molecular Weight (g/mol) | 280.17 |
MDL Number | MFCD07368291 |
InChI Key | REDKQKNJWVIPIO-UHFFFAOYSA-N |
Synonym | 4-biphenylboronic acid pinacol ester, 2-1,1'-biphenyl-4-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-4-biphenylyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-biphenylboronic acid, pinacol ester, 4,4,5,5-tetramethyl-2-4-phenylphenyl-1,3,2-dioxaborolane, 1,3,2-dioxaborolane, 2-1,1'-biphenyl-4-yl-4,4,5,5-tetramethyl, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl biphenyl, 2-biphenyl-4-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl diphenyl |
PubChem CID | 11989651 |
IUPAC Name | 4,4,5,5-tetramethyl-2-(4-phenylphenyl)-1,3,2-dioxaborolane |
SMILES | CC1(C)OB(OC1(C)C)C1=CC=C(C=C1)C1=CC=CC=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15436368
|
Thermo Scientific Alfa Aesar
H59919.03 |
1 g |
£44.70
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15446368
|
Thermo Scientific Alfa Aesar
H59919.06 |
5 g |
£188.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It is an important raw material and intermediate used in organic synthesis and pharmaceuticals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is an important raw material and intermediate used in organic synthesis and pharmaceuticals.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Chemical Identifiers
144432-80-4 | |
280.17 | |
REDKQKNJWVIPIO-UHFFFAOYSA-N | |
11989651 | |
CC1(C)OB(OC1(C)C)C1=CC=C(C=C1)C1=CC=CC=C1 |
C18H21BO2 | |
MFCD07368291 | |
4-biphenylboronic acid pinacol ester, 2-1,1'-biphenyl-4-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-4-biphenylyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-biphenylboronic acid, pinacol ester, 4,4,5,5-tetramethyl-2-4-phenylphenyl-1,3,2-dioxaborolane, 1,3,2-dioxaborolane, 2-1,1'-biphenyl-4-yl-4,4,5,5-tetramethyl, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl biphenyl, 2-biphenyl-4-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl diphenyl | |
4,4,5,5-tetramethyl-2-(4-phenylphenyl)-1,3,2-dioxaborolane |
Specifications
144432-80-4 | |
MFCD07368291 | |
Slightly soluble in water. | |
CC1(C)OB(OC1(C)C)C1=CC=C(C=C1)C1=CC=CC=C1 | |
280.17 | |
280.17 | |
Biphenyl-4-boronic acid pinacol ester |
C18H21BO2 | |
4-biphenylboronic acid pinacol ester, 2-1,1'-biphenyl-4-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-4-biphenylyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-biphenylboronic acid, pinacol ester, 4,4,5,5-tetramethyl-2-4-phenylphenyl-1,3,2-dioxaborolane, 1,3,2-dioxaborolane, 2-1,1'-biphenyl-4-yl-4,4,5,5-tetramethyl, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl biphenyl, 2-biphenyl-4-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl diphenyl | |
REDKQKNJWVIPIO-UHFFFAOYSA-N | |
4,4,5,5-tetramethyl-2-(4-phenylphenyl)-1,3,2-dioxaborolane | |
11989651 | |
97% |
Safety and Handling
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only