missing translation for 'onlineSavingsMsg'
Learn More
Biphenyl, 99%, Thermo Scientific Chemicals Product Code.: 10469010

Biphenyl, 99%, Thermo Scientific Chemicals

Product Code. 10469010
250 g, Plastic bottle
Click to view available options
Quantity:
250 g
1 kg
5 kg
Packaging:
Plastic bottle
Plastic drum
This item is not returnable. View return policy

Product Code. 10469010

Brand: Thermo Scientific Acros 106252500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 92-52-4
Molecular Formula C12H10
Molecular Weight (g/mol) 154.21
MDL Number MFCD00003054
InChI Key ZUOUZKKEUPVFJK-UHFFFAOYSA-N
Synonym biphenyl, phenylbenzene, diphenyl, 1,1'-diphenyl, bibenzene, lemonene, xenene, phenador-x, tetrosin ly, carolid al
PubChem CID 7095
ChEBI CHEBI:17097
IUPAC Name 1,1'-biphenyl
SMILES C1=CC=C(C=C1)C1=CC=CC=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 68.0°C to 72.0°C
Color White to Yellow
Boiling Point 255.0°C
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Plastic bottle
Linear Formula C6H5C6H5
Quantity 250 g
Beilstein 05, 578
Merck Index 15, 3344
Solubility Information Solubility in water: insoluble.
Formula Weight 154.21
Percent Purity 99%
Physical Form Crystalline Powder, Crystals or Flakes
Chemical Name or Material Biphenyl
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Biphenyl
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Biphenyl, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.